siRNA Details
Gene Symbol: Gcsam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159297.1 | NP_001152769.1 | 6 | 3033 | ||
NM_008099.4 | NP_032125.3 | 5 | 2967 | |||
GenBank mRNA | AK036553.1 | - | 5 | 2985 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gcsam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159297.1 | NP_001152769.1 | 6 | 3033 | ||
NM_008099.4 | NP_032125.3 | 5 | 2967 | |||
GenBank mRNA | AK036553.1 | - | 5 | 2985 |