siRNA Details
| Gene Symbol: Gpr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008154.3 | NP_032180.1 | 2 | 1385 | ||
| GenBank mRNA | AK140302.1 | - | 2 | 1356 | ||
| AK140464.1 | - | 2 | 1339 | |||
| AK158917.1 | - | 2 | 1348 | |||
| D21062.1 | - | 1 | 1759 | |||
More Information
Set Membership: |
|

Gpr3
Genomic Map