siRNA Details
| Gene Symbol: Rhpn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163465.1 | NP_001156937.1 | 1, 2 | 195 | ||
| NM_008164.2 | NP_032190.2 | 1, 2 | 195 | |||
| GenBank mRNA | AK143132.1 | - | 1, 2 | 176 | ||
| AK220431.1 | - | 1, 2 | 112 | |||
| BC004843.1 | - | 1, 2 | 165 | |||
| BC052010.1 | - | 1, 2 | 154 | 5717786 | ||
| BY081201.1 | - | 1, 2 | 196 | |||
| U43194.1 | - | 1, 2 | 89 | |||

Rhpn1
Genomic Map