siRNA Details
| Gene Symbol: Cxcl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008176.3 | NP_032202.1 | 4 | 446 | ||
| GenBank mRNA | AK140312.1 | - | 1 | 1371 | ||
| BC037997.1 | - | 4 | 413 | |||
| J04596.1 | - | 4 | 380 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cxcl1| Gene Symbol: Cxcl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008176.3 | NP_032202.1 | 4 | 446 | ||
| GenBank mRNA | AK140312.1 | - | 1 | 1371 | ||
| BC037997.1 | - | 4 | 413 | |||
| J04596.1 | - | 4 | 380 | |||