siRNA Details
| Gene Symbol: Hdac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008229.2 | NP_032255.2 | 5 | 631 | ||
| GenBank mRNA | AB221663.1 | - | 5 | 423 | ||
| AK051753.1 | - | 5 | 606 | |||
| BC138517.1 | - | 5 | 603 | 8861537 | ||
| U31758.1 | - | 5 | 591 | |||

Hdac2
Genomic Map