siRNA Details
| Gene Symbol: Hdgf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008231.4 | NP_032257.3 | 3 | 525 | ||
| GenBank mRNA | AF251787.1 | - | 3 | 438 | ||
| AK005359.1 | - | 3 | 304 | |||
| AK017863.1 | - | 3 | 479 | |||
| AK029475.1 | - | 3 | 269 | |||
| AK076021.1 | - | 3 | 313 | |||
| AK185514.1 | - | 2 | 137 | |||
| BC005713.1 | - | 3 | 435 | 2609261 | ||
| BC021654.1 | - | 3 | 453 | 4163897 | ||
| D63707.1 | - | 3 | 279 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_053707, AF448810, FQ231256, AF389348, BC070943.

Hdgf
Genomic Map