siRNA Details
| Gene Symbol: Grin2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008170.2 | NP_032196.2 | 2 | 497 | ||
| GenBank mRNA | D10217.2 | - | 2 | 497 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Grin2a| Gene Symbol: Grin2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008170.2 | NP_032196.2 | 2 | 497 | ||
| GenBank mRNA | D10217.2 | - | 2 | 497 | ||