siRNA Details
| Gene Symbol: Grin2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008171.3 | NP_032197.3 | 5 | 1311 | ||
| GenBank mRNA | AK002963.1 | - | 5 | 1312 | ||
| D10651.2 | - | 2 | 548 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Grin2b| Gene Symbol: Grin2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008171.3 | NP_032197.3 | 5 | 1311 | ||
| GenBank mRNA | AK002963.1 | - | 5 | 1312 | ||
| D10651.2 | - | 2 | 548 | |||