siRNA Details
| Gene Symbol: Hdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008230.6 | NP_032256.3 | 2 | 225 | ||
| GenBank mRNA | AF109137.1 | - | 2 | 135 | ||
| AK076384.1 | - | 2 | 209 | |||
| AK088545.1 | - | 2 | 212 | |||
| AK133455.1 | - | 2 | 226 | |||
| AK150168.1 | - | 2 | 213 | |||
| AK153104.1 | - | 2 | 213 | |||
| BC052833.1 | - | 2 | 210 | 30044098 | ||
| X57437.1 | - | 2 | 190 | |||

Hdc
Genomic Map