siRNA Details
| Gene Symbol: Golga3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008146.3 | NP_032172.3 | 3 | 555 | ||
| GenBank mRNA | AK133623.1 | - | 3 | 374 | ||
| AK171773.1 | - | 2 | 2825 | |||
| BC053002.1 | - | 3 | 555 | 5709074 | ||
| D78270.2 | - | 3 | 361 | |||

Golga3
Genomic Map