siRNA Details
| Gene Symbol: Gpc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008150.2 | NP_032176.2 | 7 | 1326 | ||
| GenBank mRNA | AI892739.1 | - | 1 | 25 | ||
| BC006622.1 | - | 7 | 1326 | 3967797 | ||
| CF168390.1 | - | 4 | 349 | |||
| X83577.1 | - | 7 | 1647 | |||

Gpc4
Genomic Map