siRNA Details
| Gene Symbol: Nr3c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008173.3 | NP_032199.3 | 7 | 2182 | ||
| GenBank mRNA | AK037733.1 | - | 7 | 2063 | ||
| AK155200.1 | - | 9 | 2264 | |||
| AK156323.1 | - | 9 | 2258 | |||
| BC129912.1 | - | 8 | 2160 | 40111802 | ||
| BC129913.1 | - | 8 | 2160 | 40111803 | ||
| DQ504162.1 | - | 7 | 2166 | |||
| HM236293.1 | - | 8 | 2142 | |||
| X04435.1 | - | 8 | 2160 | |||
| X13358.1 | - | 8 | 2142 | |||
| X13359.1 | - | 8 | 2145 | |||

Nr3c1
Genomic Map