siRNA Details
Gene Symbol: SUCNR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033050.5 | NP_149039.2 | 3 | 355 | ||
GenBank mRNA | AF247785.1 | - | 3 | 318 | ||
AF348078.1 | - | 2 | 292 | |||
AK290420.1 | - | 3 | 355 | |||
BC030948.1 | - | 3 | 349 | 4594810 | ||
EU432110.1 | - | 1 | 250 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
