siRNA Details
| Gene Symbol: Gsx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008178.2 | NP_032204.1 | 2 | 1522 | ||
| GenBank mRNA | U21224.1 | - | 2 | 1532 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gsx1| Gene Symbol: Gsx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008178.2 | NP_032204.1 | 2 | 1522 | ||
| GenBank mRNA | U21224.1 | - | 2 | 1532 | ||