siRNA Details
| Gene Symbol: DMRTB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033067.2 | NP_149056.1 | 3, 4 | 1014 | ||
| GenBank mRNA | AJ291671.1 | - | 3, 4 | 941 | ||
| AK057273.1 | - | 3, 4 | 1014 | |||
| BC029566.1 | - | 3, 4 | 530 | 4836894 | ||
| KU178739.1 | - | 3, 4 | 500 | |||

DMRTB1
Genomic Map