siRNA Details
| Gene Symbol: Hars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008214.4 | NP_032240.3 | 13 | 1601 | ||
| GenBank mRNA | AK011685.1 | - | 13 | 1517 | ||
| AK088824.1 | - | 13 | 1554 | |||
| AK146922.1 | - | 13 | 1554 | |||
| AK150081.1 | - | 13 | 1520 | |||
| AK152329.1 | - | 13 | 1533 | |||
| AK166826.1 | - | 13 | 1517 | |||
| AK166892.1 | - | 13 | 1564 | |||
| AK188864.1 | - | 2 | 62 | |||
| BC020088.1 | - | 13 | 1536 | 4008479 | ||
| U39473.1 | - | 13 | 1581 | |||

Hars
Genomic Map