siRNA Details
| Gene Symbol: Foxq1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008239.4 | NP_032265.3 | 1 | 995 | ||
| GenBank mRNA | AK147202.1 | - | 1 | 1001 | ||
| BC047155.1 | - | 1 | 728 | 6489152 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Foxq1| Gene Symbol: Foxq1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008239.4 | NP_032265.3 | 1 | 995 | ||
| GenBank mRNA | AK147202.1 | - | 1 | 1001 | ||
| BC047155.1 | - | 1 | 728 | 6489152 | ||