siRNA Details
| Gene Symbol: Hnf4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008261.2 | NP_032287.2 | 1 | 221 | ||
| GenBank mRNA | AK143948.1 | - | 1 | 246 | ||
| BC039220.1 | - | 1 | 221 | 4238842 | ||
| D29015.1 | - | 1 | 148 | |||
| EF193393.1 | - | 1 | 46 | |||

Hnf4a
Genomic Map