siRNA Details
| Gene Symbol: Ifi203 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF022371.1 | - | 450 | |||
| AK169754.1 | - | 3, 4 | 384 | |||
| AK172303.1 | - | 443 | ||||
| AK172505.1 | - | 464 | ||||
| BC008167.1 | - | 492 | 3257492 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ifi203| Gene Symbol: Ifi203 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF022371.1 | - | 450 | |||
| AK169754.1 | - | 3, 4 | 384 | |||
| AK172303.1 | - | 443 | ||||
| AK172505.1 | - | 464 | ||||
| BC008167.1 | - | 492 | 3257492 | |||