siRNA Details
| Gene Symbol: Ifi204 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008329.2 | NP_032355.2 | 6 | 1330 | ||
| GenBank mRNA | BC010546.1 | - | 1308 | 4018506 | ||
| BU694586.1 | - | 2 | 252 | |||
| CT010351.1 | - | 1222 | ||||
| M31419.1 | - | 1391 | ||||

Ifi204
Genomic Map