siRNA Details
| Gene Symbol: Hgs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159328.1 | NP_001152800.1 | 3, 4 | 298 | ||
| NM_008244.3 | NP_032270.3 | 3, 4 | 298 | |||
| GenBank mRNA | AK046299.1 | - | 3, 4 | 259 | ||
| AK145031.1 | - | 3, 4 | 218 | |||
| AK166441.1 | - | 3, 4 | 272 | |||
| AK188634.1 | - | 1, 2 | 30 | |||
| BC003239.1 | - | 3, 4 | 242 | 3497966 | ||
| BY315543.1 | - | 3, 4 | 298 | |||
| D50050.1 | - | 3, 4 | 248 | |||

Hgs
Genomic Map