siRNA Details
| Gene Symbol: Hnf4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008261.2 | NP_032287.2 | 2 | 258 | ||
| GenBank mRNA | AF015275.1 | - | 2 | 180 | ||
| AK143948.1 | - | 2 | 283 | |||
| BC039220.1 | - | 2 | 258 | 4238842 | ||
| D29015.1 | - | 2 | 185 | |||
| EF193391.1 | - | 2 | 57 | |||
| EF193393.1 | - | 2 | 83 | |||
| FJ608819.1 | - | 3 | 212 | |||
| FJ608820.1 | - | 3 | 212 | |||
| FJ608821.1 | - | 3 | 212 | |||

Hnf4a
Genomic Map