siRNA Details
Gene Symbol: Htr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008313.4 | NP_032339.2 | 4, 5 | 573 | ||
GenBank mRNA | AJ011369.1 | - | 3, 4 | 363 | ||
AK141587.1 | - | 4, 5 | 517 | |||
AK141625.1 | - | 4, 5 | 574 | |||
BC132189.1 | - | 3, 4 | 402 | |||
BC132191.1 | - | 3, 4 | 402 | |||
BC145159.1 | - | 3, 4 | 402 | |||
BC145161.1 | - | 3, 4 | 402 | |||
JN418927.1 | - | 4, 5 | 504 | |||
JN418928.1 | - | 4, 5 | 490 | |||
JN418929.1 | - | 3, 4 | 363 | |||
JN418930.1 | - | 3, 4 | 379 | |||
Y09585.1 | - | 3, 4 | 363 | |||
Y09587.1 | - | 3, 4 | 363 | |||
Y09588.1 | - | 3, 4 | 363 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
