siRNA Details
| Gene Symbol: Hus1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF076845.1 | - | 3 | 237 | ||
| AK046152.1 | - | 3 | 247 | |||
| AK051839.1 | - | 2 | 672 | |||
| AK088403.1 | - | 3 | 247 | |||
| AK135922.1 | - | 3 | 227 | |||
| BB611267.1 | - | 3 | 303 | |||
| BC047460.1 | - | 3 | 254 | |||
| BC061249.1 | - | 3 | 273 | 30249148 | ||
| Y16894.2 | - | 3 | 269 | |||

Hus1
Genomic Map