siRNA Details
| Gene Symbol: Il15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001254747.1 | NP_001241676.1 | 4 | 588 | ||
| NM_008357.2 | NP_032383.1 | 4 | 598 | |||
| GenBank mRNA | AK085441.1 | - | 6 | 798 | ||
| AK155616.1 | - | 4 | 1236 | |||
| BC023698.1 | - | 4 | 583 | 5345382 | ||
| DQ083236.1 | - | 1 | 92 | |||
| DQ083237.1 | - | 1 | 92 | |||
| U14332.1 | - | 4 | 557 | |||

Il15
Genomic Map