siRNA Details
| Gene Symbol: Il17ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008359.2 | NP_032385.1 | 5 | 605 | ||
| GenBank mRNA | AK036587.1 | - | 5 | 557 | ||
| AK080226.2 | - | 5 | 556 | |||
| U31993.1 | - | 5 | 593 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il17ra| Gene Symbol: Il17ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008359.2 | NP_032385.1 | 5 | 605 | ||
| GenBank mRNA | AK036587.1 | - | 5 | 557 | ||
| AK080226.2 | - | 5 | 556 | |||
| U31993.1 | - | 5 | 593 | |||