siRNA Details
| Gene Symbol: Il7r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008372.4 | NP_032398.3 | 1, 2 | 220 | ||
| GenBank mRNA | AF078906.1 | - | 1, 2 | 86 | ||
| AK040364.1 | - | 1, 2 | 203 | |||
| AK040740.1 | - | 1, 2 | 203 | |||
| AK041838.1 | - | 1, 2 | 111 | |||
| AK042264.1 | - | 1, 2 | 165 | |||
| AK154516.1 | - | 1, 2 | 180 | |||
| BC089571.1 | - | 1, 2 | 172 | 6758487 | ||
| BY776757.1 | - | 1, 2 | 221 | |||
| M29697.1 | - | 1, 2 | 133 | |||

Il7r
Genomic Map