siRNA Details
| Gene Symbol: Itgb1bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008403.4 | NP_032429.1 | 3 | 293 | ||
| GenBank mRNA | AJ001373.1 | - | 3 | 275 | ||
| AK002786.1 | - | 3 | 250 | |||
| AK004172.1 | - | 3 | 249 | |||
| AK007635.1 | - | 3 | 251 | |||
| AK075577.1 | - | 3 | 294 | |||
| AK089937.1 | - | 3 | 407 | |||
| AK147929.1 | - | 3 | 242 | |||
| BC021151.1 | - | 3 | 432 | |||
| BC028772.1 | - | 2 | 129 | 1230640 | ||

Itgb1bp1
Genomic Map