siRNA Details
| Gene Symbol: Itm2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008409.2 | NP_032435.2 | 2 | 284 | ||
| GenBank mRNA | AK014024.1 | - | 2 | 52 | ||
| AK051956.1 | - | 2 | 263 | |||
| AK135229.1 | - | 2 | 236 | |||
| AK167351.1 | - | 2 | 285 | |||
| BC031420.1 | - | 2 | 251 | 3668557 | ||
| BQ568915.1 | - | 2 | 252 | |||
| L38971.1 | - | 2 | 287 | |||

Itm2a
Genomic Map