siRNA Details
| Gene Symbol: Kif3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290805.1 | NP_001277734.1 | 3 | 478 | ||
| NM_001290806.1 | NP_001277735.1 | 3 | 478 | |||
| NM_008443.4 | NP_032469.2 | 3 | 478 | |||
| GenBank mRNA | AK147078.1 | - | 3 | 428 | ||
| AK161223.1 | - | 3 | 469 | |||
| AK161920.1 | - | 3 | 452 | |||
| AK169425.1 | - | 3 | 452 | |||
| BC023936.1 | - | 3 | 428 | |||
| BC044720.1 | - | 3 | 439 | |||
| BC052707.1 | - | 3 | 444 | 5702415 | ||
| D12645.1 | - | 3 | 454 | |||

Kif3a
Genomic Map