siRNA Details
| Gene Symbol: Kif4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008446.2 | NP_032472.2 | 1, 2 | 532 | ||
| GenBank mRNA | AK084837.1 | - | 1, 2 | 509 | ||
| AK135179.1 | - | 1, 2 | 480 | |||
| AK135430.1 | - | 1, 2 | 26 | |||
| AK171938.1 | - | 1, 2 | 446 | |||
| BC050946.1 | - | 1, 2 | 470 | 6822283 | ||
| BC054537.1 | - | 1, 2 | 454 | |||
| CF742002.1 | - | 1, 2 | 532 | |||
| D12646.1 | - | 1, 2 | 859 | |||

Kif4
Genomic Map