siRNA Details
Gene Symbol: Il12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK153729.1 | - | 3 | 917 | ||
BC116851.1 | - | 7 | 791 | |||
BC116855.1 | - | 7 | 791 | |||
M86672.1 | - | 7 | 887 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Il12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK153729.1 | - | 3 | 917 | ||
BC116851.1 | - | 7 | 791 | |||
BC116855.1 | - | 7 | 791 | |||
M86672.1 | - | 7 | 887 |