siRNA Details
| Gene Symbol: Il12rb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008354.3 | NP_032380.1 | 4 | 586 | ||
| GenBank mRNA | BC137745.1 | - | 3 | 498 | 8860764 | |
| U64199.1 | - | 4 | 587 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il12rb2| Gene Symbol: Il12rb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008354.3 | NP_032380.1 | 4 | 586 | ||
| GenBank mRNA | BC137745.1 | - | 3 | 498 | 8860764 | |
| U64199.1 | - | 4 | 587 | |||