siRNA Details
| Gene Symbol: Il18r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161842.1 | NP_001155314.1 | 4 | 609 | ||
| NM_001161843.1 | NP_001155315.1 | 4 | 481 | |||
| NM_008365.2 | NP_032391.1 | 3 | 534 | |||
| GenBank mRNA | AK087129.1 | - | 3 | 483 | ||
| AK089237.1 | - | 4 | 610 | |||
| AK170705.1 | - | 4 | 570 | |||
| BC020296.1 | - | 3 | 296 | |||
| BC023240.1 | - | 4 | 450 | 4483261 | ||
| U43673.1 | - | 3 | 534 | |||

Il18r1
Genomic Map