siRNA Details
| Gene Symbol: Inpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008384.2 | NP_032410.2 | 2 | 455 | ||
| GenBank mRNA | AK078382.1 | - | 2 | 343 | ||
| BC025072.1 | - | 2 | 200 | 4456812 | ||
| BY713450.1 | - | 2 | 456 | |||
| U27295.1 | - | 2 | 304 | |||

Inpp1
Genomic Map