siRNA Details
| Gene Symbol: Itm2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008409.2 | NP_032435.2 | 2 | 348 | ||
| GenBank mRNA | AK014024.1 | - | 2 | 116 | ||
| AK051956.1 | - | 2 | 327 | |||
| AK135229.1 | - | 2 | 300 | |||
| AK167351.1 | - | 2 | 349 | |||
| BC031420.1 | - | 2 | 315 | 3668557 | ||
| BQ568915.1 | - | 2 | 316 | |||
| L38971.1 | - | 2 | 351 | |||

Itm2a
Genomic Map