siRNA Details
| Gene Symbol: DUSP26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001305115.1 | NP_001292044.1 | 2 | 622 | ||
| NM_001305116.1 | NP_001292045.1 | 1 | 389 | |||
| NM_024025.2 | NP_076930.1 | 2 | 625 | |||
| GenBank mRNA | AB103376.1 | - | 2 | 440 | ||
| AB158288.1 | - | 2 | 502 | |||
| AB237597.1 | - | 2 | 437 | |||
| AK055704.1 | - | 2 | 499 | |||
| AY902194.1 | - | 2 | 440 | |||
| BC001613.1 | - | 2 | 440 | 3535215 | ||
| BC003115.1 | - | 11 | ||||
| BC067804.1 | - | 2 | 292 | 5262839 | ||
| BI753746.1 | - | 1 | 321 | |||
| BI834891.1 | - | 1 | 390 | |||

DUSP26
Genomic Map