siRNA Details
| Gene Symbol: Il17ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008359.2 | NP_032385.1 | 13 | 3232 | ||
| GenBank mRNA | AK010040.1 | - | 1 | 229 | ||
| AK036587.1 | - | 13 | 3181 | |||
| AK080226.2 | - | 11 | 4065 | |||
| BC060219.1 | - | 335 | ||||
| U31993.1 | - | 13 | 3220 | |||

Il17ra
Genomic Map