siRNA Details
| Gene Symbol: Ipp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008389.3 | NP_032415.2 | 8, 9 | 1690 | ||
| GenBank mRNA | AF285178.1 | - | 8, 9 | 1580 | ||
| AK131718.1 | - | 8, 9 | 1691 | |||
| X58523.1 | - | 618 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ipp| Gene Symbol: Ipp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008389.3 | NP_032415.2 | 8, 9 | 1690 | ||
| GenBank mRNA | AF285178.1 | - | 8, 9 | 1580 | ||
| AK131718.1 | - | 8, 9 | 1691 | |||
| X58523.1 | - | 618 | ||||