siRNA Details
| Gene Symbol: Klk1b8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008457.2 | NP_032483.1 | 2 | 144 | ||
| GenBank mRNA | BC031798.1 | - | 2 | 149 | 4167600 | |
| X03994.1 | - | 2 | 143 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klk1b8| Gene Symbol: Klk1b8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008457.2 | NP_032483.1 | 2 | 144 | ||
| GenBank mRNA | BC031798.1 | - | 2 | 149 | 4167600 | |
| X03994.1 | - | 2 | 143 | |||