siRNA Details
| Gene Symbol: Kpna3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008466.5 | NP_032492.1 | 5, 6 | 443 | ||
| GenBank mRNA | AF020772.1 | - | 5, 6 | 287 | ||
| AK011559.1 | - | 5, 6 | 314 | |||
| AK049373.1 | - | 5, 6 | 314 | |||
| AK161664.1 | - | 5, 6 | 371 | |||
| BC026885.1 | - | 5, 6 | 337 | 3979788 | ||
| CJ063714.1 | - | 5, 6 | 528 | |||

Kpna3
Genomic Map