siRNA Details
| Gene Symbol: Lag3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008479.2 | NP_032505.1 | 3, 4 | 854 | ||
| GenBank mRNA | BC120591.1 | - | 3, 4 | 786 | 40129514 | |
| X98113.1 | - | 3, 4 | 856 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lag3| Gene Symbol: Lag3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008479.2 | NP_032505.1 | 3, 4 | 854 | ||
| GenBank mRNA | BC120591.1 | - | 3, 4 | 786 | 40129514 | |
| X98113.1 | - | 3, 4 | 856 | |||