siRNA Details
| Gene Symbol: Ltk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008523.2 | NP_032549.2 | 3 | 406 | ||
| NM_203345.2 | NP_976220.2 | 9 | 1252 | |||
| NM_206941.1 | NP_996824.1 | 4 | 589 | |||
| NM_206942.2 | NP_996825.2 | 8 | 1069 | |||
| GenBank mRNA | BC047138.1 | - | 3 | 406 | 6389834 | |
| BC055898.1 | - | 3 | 400 | |||
| BC141299.1 | - | 8 | 1045 | 9055911 | ||
| M90470.1 | - | 9 | 1252 | |||
| X07984.1 | - | 3 | 380 | |||
| X52621.1 | - | 3 | 395 | |||

Ltk
Genomic Map