siRNA Details
| Gene Symbol: Masp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008555.2 | NP_032581.2 | 2 | 198 | ||
| GenBank mRNA | AB049755.1 | - | 2 | 211 | ||
| AK031598.1 | - | 2 | 259 | |||
| AK032944.1 | - | 2 | 205 | |||
| AK043055.1 | - | 2 | 258 | |||
| BC131637.1 | - | 2 | 198 | 40109637 | ||
| BC131638.1 | - | 2 | 198 | 40109639 | ||
| D16492.1 | - | 211 | ||||

Masp1
Genomic Map