siRNA Details
| Gene Symbol: Ktn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293635.1 | NP_001280564.1 | 2 | 447 | ||
| NM_001293636.1 | NP_001280565.1 | 3 | 572 | |||
| NM_008477.2 | NP_032503.2 | 1 | 318 | |||
| GenBank mRNA | AK040200.1 | - | 2 | 448 | ||
| BC046994.1 | - | 2 | 448 | |||
| BC066871.1 | - | 2 | 401 | |||
| BC075697.1 | - | 3 | 489 | |||
| CO431187.1 | - | 1 | 155 | |||
| L43326.1 | - | 1 | 306 | |||

Ktn1
Genomic Map