siRNA Details
| Gene Symbol: GPR61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031936.4 | NP_114142.3 | 2 | 2191 | ||
| XM_017002438.1 | XP_016857927.1 | 2 | 4524 | |||
| GenBank mRNA | AF258342.1 | - | 3 | 1827 | ||
More Information
Set Membership: |
|

GPR61
Genomic Map