siRNA Details
| Gene Symbol: MIXL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031944.2 | NP_114150.1 | 1, 2 | 458 | ||
| GenBank mRNA | AF211891.1 | - | 1, 2 | 394 | ||
| BC111974.1 | - | 1, 2 | 458 | 8327442 | ||
| BC113441.1 | - | 1, 2 | 458 | 8322493 | ||

MIXL1
Genomic Map