siRNA Details
| Gene Symbol: ZNF394 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032164.2 | NP_115540.2 | 3 | 956 | ||
| XM_017012711.1 | XP_016868200.1 | 2 | 877 | |||
| GenBank mRNA | AK022360.1 | - | 3 | 999 | ||
| AK172865.1 | - | 4 | 877 | |||
| AK222948.1 | - | 3 | 949 | |||
| AY642122.1 | - | 3 | 983 | |||
| BC017051.1 | - | 2 | 839 | |||
| BC025241.1 | - | 3 | 983 | 4641268 | ||
| BC051847.1 | - | 3 | 956 | 6007172 | ||
| L14843.1 | - | 1 | 406 | |||

ZNF394
Genomic Map