siRNA Details
Gene Symbol: Ktn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293635.1 | NP_001280564.1 | 2 | 519 | ||
NM_001293636.1 | NP_001280565.1 | 3 | 644 | |||
NM_008477.2 | NP_032503.2 | 1 | 390 | |||
GenBank mRNA | AK040200.1 | - | 2 | 520 | ||
BC046994.1 | - | 2 | 520 | |||
BC066871.1 | - | 2 | 473 | |||
BC075697.1 | - | 3 | 561 | |||
CO431187.1 | - | 1 | 227 | |||
L43326.1 | - | 1 | 378 |
