siRNA Details
Gene Symbol: Lag3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008479.2 | NP_032505.1 | 8 | 1828 | ||
GenBank mRNA | BC120591.1 | - | 8 | 1760 | 40129514 | |
X98113.1 | - | 8 | 1830 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lag3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008479.2 | NP_032505.1 | 8 | 1828 | ||
GenBank mRNA | BC120591.1 | - | 8 | 1760 | 40129514 | |
X98113.1 | - | 8 | 1830 |